(S)-(+)-α-Methoxyphenylacetic acid 98%

Supplier: Ambeed
Warning

A791171-1G A791171-5G A791171-100G A791171-10G A791171-250MG A791171-25G
103841-020EA 33 USD
103841-020 76845-268 76845-270 77255-878 77255-880 77255-882 77255-884
(S)-(+)-α-Methoxyphenylacetic acid 98%

Caution: for Research Use Only

Formula: C₉H₁₀O₃
Boiling Pt: 283.8 °C (760 mm Hg)
Melting Pt: 64…66 °C
Flash Pt: 116.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00064216
CAS Number: 26164-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR