2,2,2-Tribromoethanol 97%

Supplier: Ambeed
Warning

Synonyms: Tribromoethyl alcohol

A749399-25G A749399-5G A749399-100G A749399-10G A749399-1MG A749399-500G
103840-844EA 30 USD
103840-844 76845-074 76845-076 77255-580 77255-582 77255-584 77747-477
2,2,2-Tribromoethanol 97%
2,2,2-Tribromoethanol

Caution: for Research Use Only

Formula: Br₃CCH₂OH
Boiling Pt: 199 °C (760 mm Hg)
Melting Pt: 73…79 °C (760 mm Hg)
Flash Pt: 99.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00004671
CAS Number: 75-80-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR