1-Pentanesulfonic acid sodium salt monohydrate 99%

Supplier: Ambeed
Warning

Synonyms: Sodium 1-pentanesulfonate monohydrate, Sodium 1-pentanesulphonate monohydrate

A706978-25G A706978-5G A706978-100G
103840-676EA 20 USD
103840-676 76844-890 76844-892 77243-540
1-Pentanesulfonic acid sodium salt monohydrate 99%
1-Pentanesulphonic acid sodium salt monohydrate

Caution: for Research Use Only

Formula: C₅H₁₁NaO₃S·1H₂O
MW: 192.21
Storage Temperature: Ambient
MDL Number: MFCD00007541
CAS Number: 207605-40-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR