Triphenylene 98%

Supplier: Ambeed
Danger

Synonyms: 9,10-Benzophenanthrene, Isochrysene

A666345-1G A666345-25G A666345-5G A666345-250MG A666345-10G A666345-100G
103840-472EA 9 USD
103840-472 76844-672 76844-674 76844-676 77035-564 77573-148 77873-070
Triphenylene 98%
Triphenylene

Caution: for Research Use Only

Formula: C₁₈H₁₂
MW: 228.29 g/mol
Boiling Pt: 438
Melting Pt: 199
Storage Temperature: Ambient
MDL Number: MFCD00001108
CAS Number: 217-59-4
UN: 3077
ADR: 9 Ⅲ
Merck Index: 12,09870

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR