Iminodiacetic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,2'-Azanediyldiacetic acid
A617225-25G
A617225-1000G
A617225-100G
A617225-500G
76844-370EA
9
USD
76844-370
77005-494
77005-496
77005-498
Iminodiacetic acid 98%
Iminodiacetic acid
Caution: for Research Use Only
Formula:
C₄H₇NO₄ |
MDL Number:
MFCD00004280 CAS Number: 142-73-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...