4-Amino-2,2,6,6-tetramethylpiperidine 98%

Supplier: Ambeed
Warning

A611509-25G A611509-100G A611509-10G A611509-500G
103810-480EA 91 USD
103810-480 76844-336 77280-306 77280-308 77280-310
4-Amino-2,2,6,6-tetramethylpiperidine 98%
4-Amino-2,2,6,6-tetramethylpiperidine

Caution: for Research Use Only

Formula: C₉H₂₀N₂
Boiling Pt: 188.5 °C (760 mm Hg)
Melting Pt: 16…18 °C
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00005984
CAS Number: 36768-62-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR