Tetraoctylammonium bromide 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Tetra-n-octylammonium bromide
A408773-100G
A408773-10G
A408773-500G
A408773-25G
103838-238EA
5
USD
103838-238
76789-010
76789-012
76789-014
76841-750
Tetraoctylammonium bromide 98%
Tetraoctylammonium bromide
Caution: for Research Use Only
Formula:
C₃₂H₆₈BrN Melting Pt: 95…98 °C Flash Pt: 100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011863 CAS Number: 14866-33-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...