Benzyl methyl ether 98%

Supplier: Ambeed
Danger

A372710-25G A372710-5G
103837-532EA 7 USD
103837-532 76840-794 76840-796
Benzyl methyl ether 98%
Benzyl methyl ether

Caution: for Research Use Only

Formula: C₈H₁₀O
Boiling Pt: 174 °C (760 mm Hg)
Melting Pt: 52.6 °C
Flash Pt: 36.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00025901
CAS Number: 538-86-3
UN: 3271
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR