3-Bromophenylboronic acid 97%

Supplier: Ambeed
Warning

A367247-1G A367247-25G A367247-5G A367247-100G A367247-10G A367247-500G
103826-422EA 7 USD
103826-422 103837-432 76840-664 76840-666 76840-668 77115-362 77115-364 77115-366
3-Bromophenylboronic acid 97%
3-Bromophenylboronic acid

Caution: for Research Use Only

Formula: C₆H₆BBrO₂
Boiling Pt: 330.5 °C (760 mm Hg)
Melting Pt: 164…168 °C
Flash Pt: 153.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00239386
CAS Number: 89598-96-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR