2-Bromo-5-fluoropyridine 98%

Supplier: Ambeed
Warning

A335661-25G A335661-5G A335661-100G A335661-10G A335661-500G A335661-1G
103825-698EA 5 USD
103825-698 103836-670 76839-682 76839-684 77277-886 77277-888 77277-890 77306-492
2-Bromo-5-fluoropyridine 98%
2-Bromo-5-fluoropyridine

Caution: for Research Use Only

Formula: C₅H₃BrFN
Boiling Pt: 171.6 °C (760 mm Hg)
Melting Pt: 30…31 °C
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00234011
CAS Number: 41404-58-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR