2-Piperazinone 98%

Supplier: Ambeed
Warning

Synonyms: Piperazin-2-one

A319699-25G A319699-5G A319699-100G A319699-10G A319699-500G A319699-1G
103836-146EA 6 USD
103836-146 76838-996 76838-998 77114-708 77114-710 77114-712 77571-908
2-Piperazinone 98%
2-Piperazinone

Caution: for Research Use Only

Formula: C₄H₈N₂O
MW: 100.12 g/mol
Boiling Pt: 307.1 °C (760 mm Hg)
Melting Pt: 136…140 °C
Flash Pt: 166.3 °C
Storage Temperature: Ambient
MDL Number: MFCD01318687
CAS Number: 5625-67-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR