3-Benzyloxybenzyl bromide 98%

Supplier: Ambeed
Danger

Synonyms: 3-(Benzyloxy)benzyl bromide, 1-(Benzyloxy)-3-(bromomethyl)benzene

A305710-1G A305710-25G A305710-5G A305710-10G A305710-250MG
103814-444EA 7 USD
103814-444 103835-610 76838-308 76838-310 76838-312 77277-414 77277-416
3-Benzyloxybenzyl bromide 98%
3-Benzyloxybenzyl bromide

Caution: for Research Use Only

Formula: C₁₄H₁₃BrO
Boiling Pt: 365.6 °C (760 mm Hg)
Flash Pt: 142.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01318307
CAS Number: 1700-31-8
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR