2,6-Diaminopurine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
9H-Purine-2,6-diamine, 7H-Purine-2,6-diamine
A298525-25G
A298525-5G
A298525-100G
A298525-10G
A298525-1G
A298525-500G
103835-330EA
10
USD
103835-330
76838-028
76838-030
77289-918
77289-920
77289-922
77748-579
2,6-Diaminopurine 98%
2,6-Diaminopurine
Caution: for Research Use Only
Formula:
C₅H₆N₆ MW: 150.14 g/mol Boiling Pt: 749 °C (760 mm Hg) Melting Pt: 117…122 °C Flash Pt: 155 °C Storage Temperature: Ambient |
MDL Number:
MFCD00213668 CAS Number: 1904-98-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...