GC-FID Autosampler
Supplier: LUCIDITY SYSTEMS LLC
928272
76836-904EA 5062.02 USD
76836-904
GC-FID Autosampler
Autosamplers
GC-FID 16 place autosampler.
  • Self-aligning autosampler
  • Quick and easy syringe changing capabilities
  • 16-position turntable
The Lucidity GC-FID autosampler installs quickly on the GC-FID with no alignment required. Autosampler has a 16-place turn-table that allows for multiple samples to be processed automatically. Easy access allows for quick vial placement and contant visibility of sampling. The autosampler is one of the smallest and lowest cost options for automated sampling.