IonicX™ Portable XRF Analyzer
Supplier: THERMO SCIENTIFIC PORTABLE
XL5IONICXPKG
76836-894EA 84563 USD
76836-894
IonicX™ Portable XRF Analyzer
Spectrophotometers and Photometers Fluorescence Spectrophotometers
The Thermo Scientific™ portable IonicX™ XRF analyzer provides accurate ionic salt raw material identity verification and high sample throughput to pharmaceutical and biopharmaceutical manufacturers.
  • Portable verification of a wide range of ionic salts in seconds
  • Complies with 21 CFR Part 11 offering electronic signatures, user access control, and audit trails
  • Non-destructive testing of raw materials
  • Minimal sample preparation
  • Simple testing procedure without the need substantial operator training
  • Lightweight and small footprint provides easy portability
  • Wi-Fi enabled
  • Hot swap battery
  • Tilting, color, resistive touchscreen display
An innovative alternative to conventional bench-based analytical methods, IonicX™ delivers correct analysis of ionic salts at the point of use, in seconds, and with minimal sample preparation.
Applications - portable and precise verification of a wide range of ionic salts in seconds anywhere in the manufacturing plant - far faster than traditional benchtop techniques or wet
chemistry. Non-destructive testing of raw materials, preserving production volume and minimizing sampling time to ensure optimal throughput.
Regulatory compliance - The IonicX™ complies with 21 CFR Part 11, offering user access control, audit trails, electronic signatures, and full synchronization capabilities for proper storage of all electronic records.
Certifications: IP 54 certified (splash/dust proof).
Ordering information: For more information on available options and accessories, contact your Avantor representative.