HABITAT research Bioreactor, HABITAT photo ferment cct Cell Control Unit Package
Supplier: IKA Works
10007606
76836-704EA 13375.98 USD
76836-704
HABITAT research Bioreactor, HABITAT photo ferment cct Cell Control Unit Package
Bioreactors
HABITAT research is the smart laboratory bioreactor from IKA. As the first bioreactor with a lid stand, it ensures ergonomic working and a well-organized laboratory.
  • 4 peristaltic pumps allow easy tube loading, variable speed, bi-directional and speed feedback control, volume and flowrate control
  • Includes user-friendly software with functional modes like batch, fed-batch, perfusion, chaotic mixing, adjustable PID
  • New chaotic mixing option ensures a more homogeneous mixture
  • Minimal footprint to save working space
To operate HABITAT research, you only need a control unit package and a vessel package. For sensitive cell cultures such as mammalian cells, select the cell variant, with gassing rates from 0 to 2000 cc/min. If you are using HABITAT as a fermenter, we recommend the ferment variant, with gassing rates of 0 to 20000 cc/min. If you decide to use the photobioreactor, the photo version with LED panels is perfect for you. The suffix 'cct' in a package indicates that there are connection options for three additional sensors: turbidity, CO₂ and conductivity. The second step is to complement your control unit package with the vessel package that matches your application and working volume.
Certifications: CE.
Ordering information: 1+1 Year warranty.
Delivery information: Supplied with: HABITAT photo ferment S099, tablet with software for bioreactor, HA.pl. Peltier cooling element and 2 LED panels.