HABITAT Sample Flask Rack for HABITAT research Bioreactors
Supplier: IKA Works
20110862
76836-702EA 440 USD
76836-702
HABITAT Sample Flask Rack for HABITAT research Bioreactors
Bioreactors
With the help of this stand, you can safely store your sample bottles.
There are four positions for 250 ml bottles. Due to a slightly inclined position, the sample bottle can be completely emptied without any problems.