Transporter 5® Transfection Reagents
Supplier: KYFORA BIO LLC.
26008-1A 26008-5 26008-50
76836-652EA 71.45 USD
76836-652 76836-658 76836-650
Transporter 5® Transfection Reagents
Transfection Reagents
Transfection reagents for eukaryotic cells.
  • High transfection efficiency
  • Low cytotoxicity
  • High quality
  • Simple and flexible workflow
  • Adherent and suspension cells
  • DNA and protein delivery
Transporter 5® is a premium ready-to-use transfection reagent prepared from our popular linear polyethylenimine MAX (PEI MAX® 1 mg/ml). Once inside the endosome, Transporter 5® disrupts the vacuole and releases the genetic material into the cytoplasm. Stable complexation with DNA, efficient entry into the cell, and ability to escape the endosome makes Transporter 5® a powerful tool for transfection. Transporter 5® is compatible for a wide range of cell lines/types including the most commonly used HEK293 and CHO cells grown in adherent and suspension cultures.