Foam Brick
Supplier: Sonoco Thermosafe
FPP1000
76836-624CS 44.54 USD
76836-624
Foam Brick
Mailers and Shippers
Suitable for products that need refrigerated or frozen protection.
  • Vacuum-sealed for consistent weight and shape
  • Perfect when you need maximum surface contact to your payload
  • Available in multiple sizes to fit varied packing configurations
These semi-rigid foam bricks never change shape during freezing, thawing, or transit, enabling superior surface contact for more consistent temperature control.