ONE-WAY Orion Refrigerated Shipper
Supplier: Sonoco Thermosafe
05REF-OT
76836-622EA 557.34 USD
76836-622
ONE-WAY Orion Refrigerated Shipper
Mailers and Shippers
High-performance and durable shipper provides 120 hours of refrigerated (2 to 8 °C) control. Robust EPP for maximum lifespan.
  • Environmentally friendly BioPCM
  • Low payload risks
  • Refrigerated (2 to 8 °C)
The Orion refrigerated shipper is a smart box which has several recognizable and appreciated differentiators. It utilizes vacuum insulation panels which provide the highest insulating value, while using the least amount of space, to expand the available payload volume.