(3-((2-Methoxyethoxy)methyl)phenyl)boronic acid 96%

Supplier: Ambeed
Warning

A834564-100MG A834564-1G A834564-250MG A834564-5G
76832-522EA 47 USD
76832-522 76832-524 76832-526 77886-826
(3-((2-Methoxyethoxy)methyl)phenyl)boronic acid 96%

Caution: for Research Use Only

Formula: C₁₀H₁₅BO₄
MDL Number: MFCD14687274
CAS Number: 1256358-61-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR