L-Tyrosine benzyl ester 95%

Supplier: Ambeed
Warning

Synonyms: H-Tyr-OBzl, Benzyl tyrosinate, (S)-Benzyl 2-amino-3-(4-hydroxyphenyl)propanoate, (S)-2-Amino-3-(4-hydroxyphenyl)propionic acid benzyl ester

A212289-100G A212289-10G A212289-1G A212289-25G A212289-5G
103818-186EA 21 USD
103818-186 103848-884 76825-930 76825-932 77087-350 77087-352 77087-354
L-Tyrosine benzyl ester 95%
L-Tyrosine benzyl ester

Caution: for Research Use Only

Formula: C₁₆H₁₇NO₃
MW: 271.316 g/mol
Boiling Pt: 438.7 °C (760 mm Hg)
Flash Pt: 219.1 °C
Storage Temperature: Freezer
MDL Number: MFCD00191034
CAS Number: 42406-77-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR