CHIR99021 98%, Ambeed.Inc

Supplier: Ambeed
Warning

A133052-10MG A133052-1MG A133052-250MG A133052-5MG A133052-100MG A133052-1G A133052-50MG
76823-110EA 34 USD
76823-110 76823-112 76823-114 76823-116 77683-178 77683-179 77683-180
CHIR99021 98%, Ambeed.Inc
CHIR99021

Caution: For research use only.

Formula: C₂₂H₁₈Cl₂N₈
MDL Number: MFCD11846251
CAS Number: 252917-06-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR