Quercetin dihydrate 95%

Supplier: Ambeed
Danger

A114540-10G A114540-5G
76822-054EA 10 USD
76822-054 76822-056
Quercetin dihydrate 95%
Quercetin dihydrate

Caution: for Research Use Only

Formula: C₁₅H₁₄O₉
MDL Number: MFCD00149487
CAS Number: 6151-25-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR