Boc-D-His(Tos)-OH 98%

Supplier: Ambeed
Warning

A979236-1G A979236-250MG A979236-5G
76821-112EA 43 USD
76821-112 76821-114 76821-116
Boc-D-His(Tos)-OH 98%
Boc-D-His(Tos)-OH

Caution: for Research Use Only

Formula: C₁₈H₂₃N₃O₆S
MDL Number: MFCD00037890
CAS Number: 69541-68-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR