D-(+)-Glucosamine hydrochloride 98%

Supplier: Ambeed
Warning

A474862-1000G A474862-100G A474862-25G A474862-500G A474862-50MG
76818-570EA 59 USD
76818-570 76818-572 76818-574 76818-576 76818-578
D-(+)-Glucosamine hydrochloride 98%
D-(+)-Glucosamine hydrochloride

Caution: for Research Use Only

Formula: C₆H₁₃NO₅
MDL Number: MFCD00135831
CAS Number: 66-84-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR