3-Bromo-4-fluoroaniline 98%

Supplier: Ambeed
Warning

A159788-100G A159788-10G A159788-25G A159788-5G A159788-500G
103843-514EA 5 USD
103843-514 76812-658 76812-660 77319-418 77319-420 77883-253
3-Bromo-4-fluoroaniline 98%
3-Bromo-4-fluoroaniline

Caution: for Research Use Only

Formula: C₆H₅BrFN
MW: 190.01 g/mol
Boiling Pt: 235 °C (760 mm Hg)
Melting Pt: 34…36 °C
Storage Temperature: Ambient
MDL Number: MFCD03788559
CAS Number: 656-64-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR