Fmoc-Val-Cit-PAB-PNP 98%

Supplier: Ambeed
Warning

A157658-100MG A157658-10MG A157658-1G A157658-1MG A157658-250MG A157658-50MG A157658-5MG A157658-5G
76812-530EA 43 USD
76812-530 76812-532 76812-534 76812-536 76812-538 76812-540 76812-542 77883-241
Fmoc-Val-Cit-PAB-PNP 98%
Fmoc-Val-Cit-PAB-PNP

Caution: for Research Use Only

Formula: C₄₀H₄₂N₆O₁₀
MDL Number: MFCD22200278
CAS Number: 863971-53-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR