Fmoc-Val-Cit-PAB-PNP 98%
Supplier: Ambeed
|
Warning
|
A157658-100MG
A157658-10MG
A157658-1G
A157658-1MG
A157658-250MG
A157658-50MG
A157658-5MG
A157658-5G
76812-530EA
43
USD
76812-530
76812-532
76812-534
76812-536
76812-538
76812-540
76812-542
77883-241
Fmoc-Val-Cit-PAB-PNP 98%
Fmoc-Val-Cit-PAB-PNP
Caution: for Research Use Only
Formula:
C₄₀H₄₂N₆O₁₀ |
MDL Number:
MFCD22200278 CAS Number: 863971-53-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...