2-Amino-5-chlorobenzophenone 98%
Supplier: Ambeed
|
Warning
|
A153038-100G
A153038-500G
A153038-25G
76812-306EA
27
USD
76812-306
76812-308
77299-992
2-Amino-5-chlorobenzophenone 98%
2-Amino-5-chlorobenzophenone
Caution: for Research Use Only
Formula:
C₁₃H₁₀ClNO |
MDL Number:
MFCD00007839 CAS Number: 719-59-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...