ω-Bromo-3-fluoroacetophenone 97%

Supplier: Ambeed
Warning

A140366-10G A140366-1G A140366-5G A140366-100G A140366-25G
76811-738EA 39 USD
76811-738 76811-740 76811-742 77883-122 77883-123
ω-Bromo-3-fluoroacetophenone 97%
ω-Bromo-3-fluoroacetophenone

Caution: for Research Use Only

Formula: C₈H₆BrFO
MDL Number: MFCD00109756
CAS Number: 53631-18-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR