2-Indolinone 98%

Supplier: Ambeed
Warning

Synonyms: 2-Oxindole, Oxindole, Indolin-2-one, 2,3-Dihydro-2-indolone

A136297-100G A136297-10G A136297-1G A136297-500G A136297-25G A136297-5G
103833-256EA 6 USD
103833-256 76811-522 76811-524 76811-526 76811-528 77315-176 77315-178
2-Indolinone 98%
2-Indolinone

Caution: for Research Use Only

Formula: C₈H₇NO
MW: 133.15 g/mol
Boiling Pt: 313
Melting Pt: 123…128 °C
Storage Temperature: Ambient
MDL Number: MFCD00005711
CAS Number: 59-48-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR