3-Hydroxy-p-anisaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: Isovanillin, Isovanicaline, 3-Hydroxy-4-methoxybenzaldehyde, Isovanillic aldehyde

A122974-100G A122974-10G A122974-25G A122974-500G A122974-5G A122974-1000G
103831-150EA 5 USD
103831-150 76810-622 76810-624 76810-626 76810-628 77297-906 77882-991
3-Hydroxy-p-anisaldehyde 98%
3-Hydroxy-p-anisaldehyde

Caution: for Research Use Only

Formula: C₈H₈O₃
Boiling Pt: 308.1 °C (760 mm Hg)
Melting Pt: 119.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00003369
CAS Number: 621-59-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR