Undercounter Refrigerator, UR48B
Supplier: Nordon/Hoshizaki Llc.
UR48B
76809-008EA 8497.6 USD
76809-008
Undercounter Refrigerator, UR48B
Refrigerators
Streamline work with the Hoshizaki UR48B 48" undercounter refrigerator with 2 doors.
  • Includes 2 epoxy coated wire shelves adjustable in ¹/₂″ increments on anodized aluminum supports
  • Suppled as standard with 4×4″ stem casters (2 with brakes)
  • Field reversible, spring assisted self closing door with stay open feature (no kit required)
  • Electrical consumption of 1.05 kWh/day at 3 A
  • 6 years parts warranty, 5 years labor warranty and 7 years compressor warranty
  • 45 to 100 °F ambient temperature range
The UR48B Steelheart Series undercounter two section refrigerator with stainless doors is a space saving workhorse. Engineered to maintain food safe temperatures in 100 °F ambient conditions. With a net capacity of 12 cu. ft. this efficient undercounter refrigerator with front air flow design is perfect for hot/tight spaces. The UR48B is constructed of stainless steel, which protects this unit from accidental damage and rust, plus, it is easy to clean.
Certifications: The unit is ETL Safety (US & C) Listed, ETL Sanitation, Georgia Made Certified, ENERGY STAR® Qualified.
Delivery information: Supplied with 10' long cord with a 5 - 15P NEMA plug. Optional accessories such as caster kits, labels, pans and cutting board must be ordered separately.