Undercounter Refrigerator, UR27B
Supplier: Nordon/Hoshizaki Llc.
UR27B
76809-006EA 5784.37 USD
76809-006
Undercounter Refrigerator, UR27B
Refrigerators
The UR27B Steelheart Series undercounter single section refrigerator with single stainless door is a space saving workhorse.
  • Environmentally friendly R290 hydrocarbon refrigerant
  • Cabinet and doors are insulated with 2” CFC free, foamed in place polyurethane
  • Exclusive 'stepped' door design to protect door gasket
  • E-coated evaporator coil
  • Spring assisted self-closing doors with stay open feature
  • Extruded aluminum flush mount door handle
  • 45 to 100 °F ambient temperature range
Engineered to maintain food safe temperatures in 100 °F ambient conditions. With a net capacity of 6.21 cu. ft. this efficient undercounter refrigerator and front air flow design is perfect for hot/tight spaces. The UR27B is constructed of stainless steel, which protects this unit from accidental damage and rust, plus, it is easy to clean.
Certifications: The unit is ETL Safety (US & C) Listed, ETL Sanitation, Georgia Made Certified, ENERGY STAR® Qualified.
Delivery information: Supplied with 10' long cord with a 5 - 15P NEMA plug. Optional accessories such as stainless steel shelves, stacking kit (HS-5112) and 6” casters must be ordered separately.