Undercounter Freezer, UF48B
Supplier: Nordon/Hoshizaki Llc.
UF48B
76809-004EA 9552.73 USD
76809-004
Undercounter Freezer, UF48B
Freezers Laboratory / General Purpose Freezers
Increase the frozen storage capacity with the Hoshizaki Steelheart B Series UF48B 48" undercounter freezer. Made with a durable stainless steel exterior and interior, this two section undercounter unit is built to withstand tough, busy environments with ease.
  • Environmentally friendly R290 hydrocarbon refrigerant
  • Cabinet and doors are insulated with 2” CFC free, foamed in place polyurethane
  • Exclusive 'stepped' door design to protect door gasket
  • Spring assisted self-closing doors with stay open feature
  • Supplied as standard with 4” casters (two with brakes)
  • Extruded aluminum flush mount door handle
  • Anodized aluminum shelf supports adjustable in ¹/₂” increments
  • 45 to 100 °F ambient temperature range
This freezer is tested to perform in ambient temperatures of 100 degrees Fahrenheit and consistently maintains temperatures between −10 and +10 °F for efficient operation. This model has an innovative air flow design that helps to ensure even cooling in every part of the cabinet. The cabinet and doors have 2" thick CFC-free, foamed-in-place polyurethane insulation, and the ¹/₃ hp refrigeration system runs on eco-friendly R290 refrigerant, making it an environmentally responsible choice for your business.
Certifications: The unit is ETL Safety (US & C) Listed, ETL Sanitation, Georgia Made Certified, ENERGY STAR® Qualified.
Delivery information: Supplied with 10' long cord with a 5 - 15P NEMA plug. Optional accessories such as caster kits, labels, pans and stacking kit must be ordered separately.