Undercounter Freezer , UF27B
Supplier: Nordon/Hoshizaki Llc.
UF27B
76809-002EA 6698.36 USD
76809-002
Undercounter Freezer , UF27B
Freezers Laboratory / General Purpose Freezers
Streamline work with the Hoshizaki UF27B 27" undercounter freezer with 1 door.
  • Environmentally friendly R290 hydrocarbon refrigerant
  • Cabinet and doors are insulated with 2” CFC free, foamed in place polyurethane
  • Exclusive 'stepped' door design to protect door gasket
  • Spring assisted self-closing doors with stay open feature
  • Magnetic door gasket is easily removable for cleaning
  • Extruded aluminum flush mount door handle
  • Anodized aluminum shelf supports adjustable in ¹/₂” increments
  • 45 to 100 °F ambient temperature range
The UF27B Steelheart Series undercounter single section freezer with a stainless door is a space saving workhorse. Engineered to maintain food safe temperatures in 100 °F ambient conditions. With a net capacity of 6.21 cu. ft. this efficient undercounter freezer and front air flow design is perfect for hot/tight spaces. The UF27B is constructed of stainless steel, which protects this unit from accidental damage and rust, plus, it is easy to clean.
Certifications: The unit is ETL Safety (US & C) Listed, ETL Sanitation, Georgia Made Certified, ENERGY STAR® Qualified.
Delivery information: Supplied with 10' long cord with a 5 - 15P NEMA plug. Optional accessories such as caster kits, overshelves and additional epoxy shelves must be ordered separately.