MultiPlate Genie Microplate Shakers
Supplier: Scientific Industries
SI-4100
76808-694EA 1015.82 USD
76808-694
MultiPlate Genie Microplate Shakers
Shakers and Mixers Microplate Shakers
Fully programmable microplate shaker for four plates (8 with optional tray).
  • Provides consistent well-to-well, plate-to-plate mixing for 96 and 384-well microplates, deep well plates, or PCR plates
  • Accepts PCR work up racks with standard microplate footprint for use with PCR tubes and strips
  • Featuring programmable speed, time, pulsing/interval mixing - for accurate and repeatable results
Digital true vortex mixing microplate shaker handles up to 4 microplates, (or up to 8 plates with the optional accessory tray). The combined speed and 1 mm orbit provide consistant well to well and plate to plate uniformity, even in 384-well plates. Featuring programable speed, time and continuous or pulse time, it's ideal for achieving accurate and repeatable results. The Pulse feature allows rapid pulsing or timed 'on/off' mixing for interval mixing. Ideal for use in PCR preparation, digestions, bead mixing, ELISA, EIA, and colorimetric assay applications.
Pulse (Interval): 0.1 s to 99 min
Certifications: CE marked.
Delivery information: Supplied with tray for 4 microplates. An optional stacking tray accessory for additional 4 microplates (total of 8) is available to purchase separately.