Rapid Clear® Cap 3000, Thomson Instrument Company
Supplier: THOMSON INSTRUMENT CO
788116
103507-532CS 995 USD
103507-532 76808-350
Rapid Clear® Cap 3000, Thomson Instrument Company
Closures Bottle Closures
Designed for high-speed clarification of 2 to 4 litres of cell culture.
  • Save time by clarifying 3 litres of cell culture in less than 35 minutes
  • 0.2 μm filter eliminates multiple filtration steps, and in some cases centrifugation
  • Cell clarification of low or high density cultures of CHO stable, CHO transient, HEK293, hybridoma, and other mammalian cell lines
  • Eliminate transfer steps: The Rapid Clear® Cap screws directly onto Optimum Growth™ Flasks
Use with 2.8 and 5 litres Optimum Growth™ Flasks.