Hexaamminecobalt trichloride 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Hexaamminecobalt(iii)chloride, Hexammine cobalt(III) chloride, Hexaamminecobalt (III) chloride
A973623-25G
A973623-5G
76808-174EA
99
USD
76808-174
77603-372
Hexaamminecobalt trichloride 98%
Hexaamminecobalt trichloride
Caution: for Research Use Only
Formula:
H₁₈CoCl₃N₆ |
MDL Number:
MFCD00036304 CAS Number: 10534-89-1 UN: 2811 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...