Lithium benzoate 99%

Supplier: Ambeed
Warning

Synonyms: Benzoic acid lithium salt

A771737-1000G A771737-100G A771737-500G A771737-25G A771737-5G
76807-310EA 299 USD
76807-310 76807-312 76807-314 77569-466 77880-547
Lithium benzoate 99%
Lithium benzoate

Caution: For research use only.

Formula: C₇H₅LiO₂
MDL Number: MFCD00035540
CAS Number: 553-54-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR