Ethidium bromide 95%

Supplier: Ambeed
Danger

A752777-1G
76807-162EA 22 USD
76807-162
Ethidium bromide 95%
Ethidium bromide

Caution: for Research Use Only

Formula: C₂₁H₂₀BrN₃
MDL Number: MFCD00011724
CAS Number: 1239-45-8
UN: 2811
ADR: 6.1,Ⅰ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR