Magnesium acetylacetonate 98%

Supplier: Ambeed
Warning

Synonyms: Magnesium bis(2,4-pentanedionato-O,O'), Bis(2,4-pentanedionato-O,O')magnesium, Magnesiumacetylacetonate, Magnesium 2,4-pentanedionate

A747755-1G A747755-5G
76807-130EA 15 USD
76807-130 76807-132
Magnesium acetylacetonate 98%
Magnesium acetylacetonate

Caution: for Research Use Only

Formula: C₁₀H₁₄MgO₄
MDL Number: MFCD00013497
CAS Number: 14024-56-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR