Lithium bis(trifluoromethylsulfonyl)imide 98%

Supplier: Ambeed
Danger

Synonyms: Lithium Bis(Trifluoromethanesulfonyl)Imide, Bis(trifluoromethane)sulfonimide lithium salt, Lithium bis(trifluoromethanesulphonimide), Lithium bis((trifluoromethyl)sulphonyl)amide, Lithium bis((trifluoromethyl)sulfonyl)amide, Lithium bis(trifluoromethanesulfonimide), Lithium Bis(Trifluoromethanesulphonyl)Imide, Bis(trifluoromethane)sulphonimide lithium salt

A725292-10G A725292-25G A725292-5G
76806-942EA 20 USD
76806-942 76806-944 76806-946
Lithium bis(trifluoromethylsulfonyl)imide 98%
Lithium bis(trifluoromethylsulfonyl)imide

Caution: for Research Use Only

Formula: C₂F₆LiNO₄S₂
MW: 287.09 g/mol
MDL Number: MFCD00210017
CAS Number: 90076-65-6
UN: 2923
ADR: 8,6.1 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR