(1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl-(S)-2-hydroxypropionate 98%

Supplier: Ambeed
Danger

Synonyms: (S)-2-Hydroxypropionic acid (1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl Ester, (S)-(1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl 2-hydroxypropanoate, L-Menthyl L-Lactate

A707629-5G
76806-772EA 9.55 USD
76806-772
(1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl-(S)-2-hydroxypropionate 98%
(1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl-(S)-2-hydroxypropionate

Caution: for Research Use Only

Formula: C₁₃H₂₄O₃
MW: 228.33 g/mol
MDL Number: MFCD27977194
CAS Number: 61597-98-6
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR