Diosmin 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-((((2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one
A670506-100G
A670506-1G
A670506-25G
A670506-500G
A670506-50MG
A670506-5G
76806-542EA
82
USD
76806-542
76806-544
76806-546
76806-548
76806-550
76806-552
Diosmin 95%
Diosmin
Caution: for Research Use Only
Formula:
C₂₈H₃₂O₁₅ MW: 608.5 g/mol |
MDL Number:
MFCD00009772 CAS Number: 520-27-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...