Diosmin 95%

Supplier: Ambeed
Warning

Synonyms: 5-Hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-((((2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one

A670506-100G A670506-1G A670506-25G A670506-500G A670506-50MG A670506-5G
76806-542EA 82 USD
76806-542 76806-544 76806-546 76806-548 76806-550 76806-552
Diosmin 95%
Diosmin

Caution: for Research Use Only

Formula: C₂₈H₃₂O₁₅
MW: 608.5 g/mol
MDL Number: MFCD00009772
CAS Number: 520-27-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR