3-(Trifluoromethyl)phenylacetone 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-[3-(Trifluoromethyl)phenyl]-2-propanone
A633756-1G
A633756-5G
A633756-25G
76806-228EA
10
USD
76806-228
76806-230
77880-342
3-(Trifluoromethyl)phenylacetone 95%
3-(Trifluoromethyl)phenylacetone
Caution: for Research Use Only
Formula:
C₁₀H₉F₃O |
MDL Number:
MFCD00000397 CAS Number: 21906-39-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...