3-(Trifluoromethyl)phenylacetone 95%

Supplier: Ambeed
Warning

Synonyms: 1-[3-(Trifluoromethyl)phenyl]-2-propanone

A633756-1G A633756-5G A633756-25G
76806-228EA 10 USD
76806-228 76806-230 77880-342
3-(Trifluoromethyl)phenylacetone 95%
3-(Trifluoromethyl)phenylacetone

Caution: for Research Use Only

Formula: C₁₀H₉F₃O
MDL Number: MFCD00000397
CAS Number: 21906-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR