di-Lithium tetraborate 98%

Supplier: Ambeed
Danger

A610844-25G
76806-062EA 15 USD
76806-062
di-Lithium tetraborate 98%
di-Lithium tetraborate

Caution: for Research Use Only

Formula: Li₂B₄O₇
MDL Number: MFCD00011083
CAS Number: 12007-60-2
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR